Product Details

SNP ID
rs12663184
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:30333823 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATGTGCCTATTTCCAAAATATCCT[C/T]AGAGTGTGTTAACAGCTTCATCAAG
Phenotype
MIM: 605700
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HCG17 PubMed Links
Additional Information
For this assay, SNP(s) [rs17194852] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HCG17
Gene Name
HLA complex group 17 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HCG18
Gene Name
HLA complex group 18 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TRIM39
Gene Name
tripartite motif containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021253.3 Intron NP_067076.2
NM_172016.2 Intron NP_742013.1
Gene
TRIM39-RPP21
Gene Name
TRIM39-RPP21 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199119.1 Intron NP_001186048.1

View Full Product Details