Product Details
- SNP ID
-
rs6663474
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:23873152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATTCCTCTGCAATCTTCCTGCC[A/G]CCAACAATTTCTCTTGTTGGCTGTT
- Phenotype
-
MIM: 605051
MIM: 612280
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs189596747] are located under a probe and SNP(s) [rs74063324] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNR2
- Gene Name
- cannabinoid receptor 2
- Gene
- FUCA1
- Gene Name
- fucosidase, alpha-L- 1, tissue
There are no transcripts associated with this gene.
View Full Product Details