Product Details

SNP ID
rs11614224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120454433 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTTGAGAGCCTAAGCTCTTAGC[A/C]TTTTTGAGACGGAGTCTTGCTCTGT
Phenotype
MIM: 601943 MIM: 614943
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GATC PubMed Links

Gene Details

Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176818.2 Intron NP_789788.1
Gene
SRSF9
Gene Name
serine and arginine rich splicing factor 9
There are no transcripts associated with this gene.

Gene
TRIAP1
Gene Name
TP53 regulated inhibitor of apoptosis 1
There are no transcripts associated with this gene.

View Full Product Details