Product Details

SNP ID
rs11855354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78151289 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTCTGTTGAGTTTCCATTTCAG[A/G]TTGTGGCCAGGCATGGTGGCTCATG
Phenotype
MIM: 601149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDH3A PubMed Links

Gene Details

Gene
IDH3A
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005530.2 Intron NP_005521.1
XM_005254336.2 Intron XP_005254393.1
XM_017022118.1 Intron XP_016877607.1
XM_017022119.1 Intron XP_016877608.1
XM_017022120.1 Intron XP_016877609.1

View Full Product Details