Product Details

SNP ID
rs8113682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19632921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACCCTTCACCTGATCTCCATGT[G/T]GCCAGCTCACTCCTTCATGTCCTTC
Phenotype
MIM: 609694 MIM: 605110
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GMIP PubMed Links

Gene Details

Gene
GMIP
Gene Name
GEM interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288998.1 Intron NP_001275927.1
NM_001288999.1 Intron NP_001275928.1
NM_016573.3 Intron NP_057657.2
XM_005259927.2 Intron XP_005259984.1
XM_011528048.2 Intron XP_011526350.1
XM_017026861.1 Intron XP_016882350.1
XM_017026862.1 Intron XP_016882351.1
Gene
LPAR2
Gene Name
lysophosphatidic acid receptor 2
There are no transcripts associated with this gene.

View Full Product Details