Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033925.1 | 1502 | Silent Mutation | CAA,CAG | Q309Q | NP_001029097.1 |
NM_001323964.1 | 1502 | Silent Mutation | CAA,CAG | Q169Q | NP_001310893.1 |
NM_001323965.1 | 1502 | Silent Mutation | CAA,CAG | Q169Q | NP_001310894.1 |
NM_001323967.1 | 1502 | Silent Mutation | CAA,CAG | Q169Q | NP_001310896.1 |
NM_001323968.1 | 1502 | Silent Mutation | CAA,CAG | Q270Q | NP_001310897.1 |
NM_001323969.1 | 1502 | Silent Mutation | CAA,CAG | Q253Q | NP_001310898.1 |
NM_001323970.1 | 1502 | Silent Mutation | CAA,CAG | Q270Q | NP_001310899.1 |
NM_003252.3 | 1502 | Silent Mutation | CAA,CAG | Q292Q | NP_003243.1 |
XM_017016605.1 | 1502 | Intron | XP_016872094.1 | ||
XM_017016606.1 | 1502 | Silent Mutation | CAA,CAG | Q169Q | XP_016872095.1 |