Product Details

SNP ID
rs1065369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141738934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTCAACACAAGGTGGTCAGGGT[A/G]AAGGCCGTGATTCAGCAGTGAACTT
Phenotype
MIM: 603863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF7 PubMed Links

Gene Details

Gene
RNF7
Gene Name
ring finger protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201370.1 Intron NP_001188299.1
NM_014245.4 Intron NP_055060.1
NM_183237.2 Intron NP_899060.1

View Full Product Details