Product Details

SNP ID
rs1401570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186108667 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATCTCTCTGTCTTCACCTCACT[G/T]TCCTGCATCCCTGGATACGTGATCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM149A PubMed Links

Gene Details

Gene
FAM149A
Gene Name
family with sequence similarity 149 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006655.2 Intron NP_001006656.1
NM_015398.2 Intron NP_056213.1
XM_005262911.1 Intron XP_005262968.1
XM_005262912.1 Intron XP_005262969.1
XM_005262913.1 Intron XP_005262970.1
XM_011531834.1 Intron XP_011530136.1
XM_011531835.2 Intron XP_011530137.1
XM_011531836.1 Intron XP_011530138.1
XM_017007996.1 Intron XP_016863485.1
XM_017007997.1 Intron XP_016863486.1
XM_017007998.1 Intron XP_016863487.1
XM_017007999.1 Intron XP_016863488.1
XM_017008000.1 Intron XP_016863489.1
XM_017008001.1 Intron XP_016863490.1
XM_017008002.1 Intron XP_016863491.1
XM_017008003.1 Intron XP_016863492.1
XM_017008004.1 Intron XP_016863493.1
XM_017008005.1 Intron XP_016863494.1
XM_017008006.1 Intron XP_016863495.1
XM_017008007.1 Intron XP_016863496.1
XM_017008008.1 Intron XP_016863497.1

View Full Product Details