Product Details

SNP ID
rs937861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69094080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAAAGTATCCTCTCTGCCTGAT[G/T]CACCTCACCCTGAGATGGCAAAAGG
Phenotype
MIM: 605709
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL6IP5 PubMed Links

Gene Details

Gene
ARL6IP5
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006407.3 Intron NP_006398.1

View Full Product Details