Product Details

SNP ID
rs1063602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:863830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCCAGGCTCATCAATTAAACA[A/G]AAAACAGGGGAGCTCTCCTCACCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD9 PubMed Links

Gene Details

Gene
BRD9
Gene Name
bromodomain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009877.2 2426 UTR 3 NP_001009877.2
NM_001317951.1 2426 UTR 3 NP_001304880.1
NM_023924.4 2426 UTR 3 NP_076413.3
XM_006714494.3 2426 Intron XP_006714557.1
XM_017009780.1 2426 Intron XP_016865269.1
Gene
ZDHHC11
Gene Name
zinc finger DHHC-type containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024786.2 2426 Intron NP_079062.1
XM_017009869.1 2426 Intron XP_016865358.1
XM_017009870.1 2426 Intron XP_016865359.1
XM_017009871.1 2426 Intron XP_016865360.1
XM_017009872.1 2426 Intron XP_016865361.1
XM_017009873.1 2426 Intron XP_016865362.1
XM_017009874.1 2426 Intron XP_016865363.1
XM_017009875.1 2426 Intron XP_016865364.1
XM_017009876.1 2426 Intron XP_016865365.1
XM_017009877.1 2426 Intron XP_016865366.1
XM_017009878.1 2426 Intron XP_016865367.1

View Full Product Details