Product Details

SNP ID
rs1188868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138908606 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACGCCTGGGCCAGTTTGATTA[C/T]TTTTAATTGTCGTGCTTTTTCTTCC
Phenotype
MIM: 614825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECT2L PubMed Links

Gene Details

Gene
ECT2L
Gene Name
epithelial cell transforming 2 like
There are no transcripts associated with this gene.

Gene
REPS1
Gene Name
RALBP1 associated Eps domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128617.2 Intron NP_001122089.1
NM_001286611.1 Intron NP_001273540.1
NM_001286612.1 Intron NP_001273541.1
NM_031922.4 Intron NP_114128.3
XM_005267177.3 Intron XP_005267234.1
XM_005267178.4 Intron XP_005267235.1
XM_005267179.3 Intron XP_005267236.1
XM_011536202.2 Intron XP_011534504.1
XM_017011387.1 Intron XP_016866876.1
XM_017011388.1 Intron XP_016866877.1
XM_017011389.1 Intron XP_016866878.1

View Full Product Details