Product Details
- SNP ID
-
rs1681938
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:63642700 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTATGTTTTAATTATTTAGAATGCC[A/G]GTAGATGGGCTTCCTATGTTTTATT
- Phenotype
-
MIM: 607789
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PHF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115241979,rs78318979] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHF3
- Gene Name
- PHD finger protein 3
View Full Product Details