Product Details

SNP ID
rs806507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46551748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGCACCTGACAACAGAGTGTT[C/T]AGGACATGCTTTGACAACCCATCCC
Phenotype
MIM: 605994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP39A1 PubMed Links

Gene Details

Gene
CYP39A1
Gene Name
cytochrome P450 family 39 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278738.1 Intron NP_001265667.1
NM_001278739.1 Intron NP_001265668.1
NM_016593.4 Intron NP_057677.2
XM_005249171.3 Intron XP_005249228.1
XM_011514658.1 Intron XP_011512960.1
XM_017010921.1 Intron XP_016866410.1
XM_017010922.1 Intron XP_016866411.1
XM_017010923.1 Intron XP_016866412.1
XM_017010924.1 Intron XP_016866413.1
XM_017010925.1 Intron XP_016866414.1

View Full Product Details