Product Details

SNP ID
rs1526090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87402493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGTTATGTTCTGCTTTTTAAAT[A/G]TAACACAAATGGGTTCCCATGCTGT
Phenotype
MIM: 171060 MIM: 606090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB4 PubMed Links

Gene Details

Gene
ABCB4
Gene Name
ATP binding cassette subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000443.3 Intron NP_000434.1
NM_018849.2 Intron NP_061337.1
NM_018850.2 Intron NP_061338.1
XM_011516308.2 Intron XP_011514610.2
XM_011516309.2 Intron XP_011514611.2
XM_011516310.2 Intron XP_011514612.2
XM_011516311.2 Intron XP_011514613.2
XM_011516312.2 Intron XP_011514614.2
XM_011516313.2 Intron XP_011514615.1
XM_011516315.2 Intron XP_011514617.2
XM_017012323.1 Intron XP_016867812.1
Gene
CROT
Gene Name
carnitine O-octanoyltransferase
There are no transcripts associated with this gene.

View Full Product Details