Product Details

SNP ID
rs1047035
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:98846664 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCCCTGCCCTTCCTGACAGGCTC[C/T]GGCTACTTCTGGCCTCATTATTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM130 PubMed Links

Gene Details

Gene
TMEM130
Gene Name
transmembrane protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134450.1 2547 UTR 3 NP_001127922.1
NM_001134451.1 2547 UTR 3 NP_001127923.1
NM_152913.2 2547 UTR 3 NP_690877.1

View Full Product Details