Product Details
- SNP ID
-
rs659939
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:54236472 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGCAGAGATGGTGAGGATGTCTT[C/G]CTTTGTTACCCAGGTTGGTCTTGAA
- Phenotype
-
MIM: 607390
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SSBP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77243666] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SSBP3
- Gene Name
- single stranded DNA binding protein 3
- Gene
- SSBP3-AS1
- Gene Name
- SSBP3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details