Product Details
- SNP ID
-
rs1261893
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:156779938 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGGAAAGAATGAGGAACTTTCTC[A/C]CACCTTTAGCCACAGCACGTTCCCT
- Phenotype
-
MIM: 614556
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARID1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117079245] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARID1B
- Gene Name
- AT-rich interaction domain 1B
- Gene
- MIR4466
- Gene Name
- microRNA 4466
There are no transcripts associated with this gene.
View Full Product Details