Product Details

SNP ID
rs1021728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99647850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTTGTGAAGTGGAATATCAGG[C/T]TCAAATGGAGGTCCCTCCCGAATGA
Phenotype
MIM: 607815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 101 Intron NP_001190994.1
NM_001204066.1 101 Intron NP_001190995.1
NM_001204067.1 101 Intron NP_001190996.1
NM_001204068.1 101 Intron NP_001190997.1
NM_001204069.1 101 Intron NP_001190998.1
NM_001204070.1 101 Intron NP_001190999.1
NM_001204079.1 101 Intron NP_001191008.1
NM_001204080.1 101 Intron NP_001191009.1
NM_001204081.1 101 Intron NP_001191010.1
NM_020140.3 101 Intron NP_064525.1
NM_152788.4 101 Intron NP_690001.3
NM_181670.3 101 Intron NP_858056.2
XM_005269028.4 101 Intron XP_005269085.1
XM_005269029.4 101 Intron XP_005269086.1
XM_005269032.3 101 Intron XP_005269089.1
XM_006719504.3 101 Intron XP_006719567.1
XM_006719505.3 101 Intron XP_006719568.1
XM_006719506.3 101 Intron XP_006719569.1
XM_006719507.3 101 Intron XP_006719570.1
XM_006719508.3 101 Intron XP_006719571.1
XM_006719509.3 101 Intron XP_006719572.1
XM_006719510.3 101 Intron XP_006719573.1
XM_006719512.3 101 Intron XP_006719575.1
XM_006719513.3 101 Intron XP_006719576.1
XM_006719514.3 101 Intron XP_006719577.1
XM_011538571.2 101 Intron XP_011536873.1
XM_017019651.1 101 Intron XP_016875140.1
XM_017019652.1 101 Intron XP_016875141.1
XM_017019653.1 101 Intron XP_016875142.1
XM_017019654.1 101 Intron XP_016875143.1
XM_017019655.1 101 Intron XP_016875144.1
XM_017019656.1 101 Intron XP_016875145.1
XM_017019657.1 101 Intron XP_016875146.1
XM_017019658.1 101 Intron XP_016875147.1
XM_017019659.1 101 Intron XP_016875148.1
XM_017019660.1 101 Intron XP_016875149.1
XM_017019661.1 101 Intron XP_016875150.1
XM_017019662.1 101 Intron XP_016875151.1
XM_017019663.1 101 Intron XP_016875152.1
XM_017019664.1 101 Intron XP_016875153.1
XM_017019665.1 101 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 101 UTR 5 NP_699195.1

View Full Product Details