Product Details

SNP ID
rs955171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127803528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTGGGGGAGAGGCAGGAATCA[G/T]GGGTAGTGGAGGGCCCCAGAACCTG
Phenotype
MIM: 136510
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FPGS PubMed Links

Gene Details

Gene
FPGS
Gene Name
folylpolyglutamate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018078.2 Intron NP_001018088.1
NM_001288803.1 Intron NP_001275732.1
NM_004957.5 Intron NP_004948.4
XM_005251864.3 Intron XP_005251921.1
XM_011518439.2 Intron XP_011516741.1
XM_017014565.1 Intron XP_016870054.1
XM_017014566.1 Intron XP_016870055.1

View Full Product Details