Product Details

SNP ID
rs9160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2732743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTCCGGCGGCAGGGACAATGGC[A/G]AGGCCGCTCACCACTTGAGGAAGAC
Phenotype
MIM: 612168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC39A3 PubMed Links

Gene Details

Gene
SLC39A3
Gene Name
solute carrier family 39 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144564.4 1207 UTR 3 NP_653165.2
NM_213568.1 1207 Intron NP_998733.1

View Full Product Details