Product Details

SNP ID
rs6630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4090424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAGAGGCACCCCGGCCAGGAC[G/T]GGCAGGAGAGGAGACCCCCGTTCCT
Phenotype
MIM: 601263
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP2K2 PubMed Links

Gene Details

Gene
MAP2K2
Gene Name
mitogen-activated protein kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030662.3 Intron NP_109587.1
XM_006722799.2 Intron XP_006722862.1
XM_017026989.1 Intron XP_016882478.1
XM_017026990.1 Intron XP_016882479.1
XM_017026991.1 Intron XP_016882480.1

View Full Product Details