Product Details

SNP ID
rs1549257
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7479702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGCCGCAGCAGGCGGGGCACAC[A/G]GGACTTCGATGGCCGTGAAACAATT
Phenotype
MIM: 616432 MIM: 607583
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARHGEF18 PubMed Links
Additional Information
For this assay, SNP(s) [rs75977782] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ARHGEF18
Gene Name
Rho/Rac guanine nucleotide exchange factor 18
There are no transcripts associated with this gene.

Gene
LOC100128573
Gene Name
uncharacterized LOC100128573
There are no transcripts associated with this gene.

Gene
PEX11G
Gene Name
peroxisomal biogenesis factor 11 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270539.1 494 Intron NP_001257468.1
NM_001300881.1 494 Intron NP_001287810.1
NM_080662.3 494 Intron NP_542393.1
XM_005272506.2 494 Intron XP_005272563.1
XM_011528426.2 494 Missense Mutation CCG,CTG P181L XP_011526728.1
XM_011528427.2 494 Missense Mutation CCG,CTG P155L XP_011526729.1
XM_011528428.1 494 Intron XP_011526730.1
XM_011528429.2 494 Intron XP_011526731.1
XM_011528431.2 494 Intron XP_011526733.1
XM_011528432.1 494 Intron XP_011526734.1

View Full Product Details