Product Details

SNP ID
rs12625573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63343330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGGCGGCCGCACCTGGGCTCGG[C/T]GGGCCACACGGTCGGCGGGGCTTGG
Phenotype
MIM: 118504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNA4 PubMed Links

Gene Details

Gene
CHRNA4
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000744.6 5523 UTR 3 NP_000735.1
NM_001256573.1 5523 UTR 3 NP_001243502.1
XM_011528524.1 5523 Intron XP_011526826.1
XM_017027624.1 5523 Intron XP_016883113.1
XM_017027625.1 5523 Intron XP_016883114.1
Gene
COL20A1
Gene Name
collagen type XX alpha 1 chain
There are no transcripts associated with this gene.

View Full Product Details