Product Details

SNP ID
rs1574103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67290128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGGCGGACCCCACTGGAGCTGGC[C/T]GTGTCTCTGGGAAACCTGGAGTCTG
Phenotype
MIM: 615126 MIM: 109635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207354.2 311 Silent Mutation GCC,GCT A47A NP_997237.2
XM_011544985.1 311 Silent Mutation GCC,GCT A47A XP_011543287.1
XM_011544986.1 311 UTR 5 XP_011543288.1
XM_011544987.2 311 UTR 5 XP_011543289.1
XM_011544988.2 311 Intron XP_011543290.1
Gene
GRK2
Gene Name
G protein-coupled receptor kinase 2
There are no transcripts associated with this gene.

View Full Product Details