Product Details

SNP ID
rs1481498
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:98894134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAATTTGGGGCATTATCTTCTA[A/G]GCTTGACCCACTTAATACTAGAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLPPR5 PubMed Links

Gene Details

Gene
PLPPR5
Gene Name
phospholipid phosphatase related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010861.2 Intron NP_001010861.1
NM_001037317.1 Intron NP_001032394.1
XM_011540836.2 Intron XP_011539138.1
XM_011540837.1 Intron XP_011539139.1
XM_011540838.2 Intron XP_011539140.1

View Full Product Details