Product Details

SNP ID
rs2330875
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:43585513 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTACAAAAGGAGTAGATTGTAAAGC[A/T]GCATAACAATGCAGTTATTAGATAA
Phenotype
MIM: 604726
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STK17A PubMed Links

Gene Details

Gene
STK17A
Gene Name
serine/threonine kinase 17a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004760.2 Intron NP_004751.2
XM_017012792.1 Intron XP_016868281.1

View Full Product Details