Product Details

SNP ID
rs405729
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:52977983 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CTCATAAATTCTTTATTTTGGCTAA[C/T]ACTGATTGCATTCATTACTACCATT
Phenotype
MIM: 605450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA4 PubMed Links

Gene Details

Gene
GSTA4
Gene Name
glutathione S-transferase alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001512.3 1306 UTR 3 NP_001503.1
XM_005249035.3 1306 UTR 3 XP_005249092.1
XM_011514534.2 1306 UTR 3 XP_011512836.1
XM_011514535.2 1306 UTR 3 XP_011512837.1

View Full Product Details