Product Details
- SNP ID
-
rs134608
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29079221 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTTCTAAGGGAATTCCTTCCCC[C/T]CCTTTTAAATTCTCTTTCTAGGTAA
- Phenotype
-
MIM: 609898
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C22orf31
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201768818] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C22orf31
- Gene Name
- chromosome 22 open reading frame 31
There are no transcripts associated with this gene.
- Gene
- KREMEN1
- Gene Name
- kringle containing transmembrane protein 1
View Full Product Details