Product Details

SNP ID
rs1059628
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:201135425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATTCCCTTCTGGCCTGCCTTC[C/T]CCCTCCCTTCAACCCCAAAGACCCA
Phenotype
MIM: 616877
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM9 PubMed Links

Gene Details

Gene
TMEM9
Gene Name
transmembrane protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288564.1 919 UTR 3 NP_001275493.1
NM_001288565.1 919 UTR 3 NP_001275494.1
NM_001288566.1 919 UTR 3 NP_001275495.1
NM_001288567.1 919 UTR 3 NP_001275496.1
NM_001288568.1 919 UTR 3 NP_001275497.1
NM_001288569.1 919 UTR 3 NP_001275498.1
NM_001288570.1 919 UTR 3 NP_001275499.1
NM_001288571.1 919 UTR 3 NP_001275500.1
NM_016456.4 919 UTR 3 NP_057540.1
XM_011509385.1 919 UTR 3 XP_011507687.1
XM_011509386.2 919 UTR 3 XP_011507688.1
XM_011509387.2 919 UTR 3 XP_011507689.1

View Full Product Details