Product Details

SNP ID
rs1380576
Assay Type
Validated
NCBI dbSNP Submissions
39
Location
Chr.1:204519150 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACGACTAAACAGTGCAAGATAAGA[C/G]TGAAAATGTGCTGAGGGGATAGACC
Phenotype
MIM: 602704
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MDM4 PubMed Links

Gene Details

Gene
MDM4
Gene Name
MDM4, p53 regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204171.1 Intron NP_001191100.1
NM_001204172.1 Intron NP_001191101.1
NM_001278516.1 Intron NP_001265445.1
NM_001278517.1 Intron NP_001265446.1
NM_001278518.1 Intron NP_001265447.1
NM_001278519.1 Intron NP_001265448.1
NM_002393.4 Intron NP_002384.2
XM_017001311.1 Intron XP_016856800.1
XM_017001312.1 Intron XP_016856801.1
XM_017001313.1 Intron XP_016856802.1

View Full Product Details