Product Details

SNP ID
rs1046925
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:205142658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTTCAAAATGCACACAATTAA[A/G]ACGGTCCTGCTGTAACAATATTATG
Phenotype
MIM: 612666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSTYK PubMed Links

Gene Details

Gene
DSTYK
Gene Name
dual serine/threonine and tyrosine protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015375.2 7685 UTR 3 NP_056190.1
NM_199462.2 7685 UTR 3 NP_955749.1
XM_011509392.2 7685 UTR 3 XP_011507694.1
XM_011509393.2 7685 UTR 3 XP_011507695.1
XM_011509394.2 7685 UTR 3 XP_011507696.1

View Full Product Details