Product Details

SNP ID
rs1342820
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:113771933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGAAAAACAAGACTTTTTTTT[A/T]AAAGAAGTTATGGTACCAAATATAA
Phenotype
MIM: 615858
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RSBN1 PubMed Links

Gene Details

Gene
RSBN1
Gene Name
round spermatid basic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018364.4 Intron NP_060834.2
XM_017001518.1 Intron XP_016857007.1

View Full Product Details