Product Details

SNP ID
rs909112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1919577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGGCCATTCCTATACTTGGTA[A/G]GGGGCCTGCACGGGCATAGCCCCCC
Phenotype
MIM: 600692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNNT3 PubMed Links

Gene Details

Gene
TNNT3
Gene Name
troponin T3, fast skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042780.2 9 UTR 5 NP_001036245.1
NM_001042781.2 9 UTR 5 NP_001036246.1
NM_001042782.2 9 UTR 5 NP_001036247.1
NM_001297646.1 9 UTR 5 NP_001284575.1
NM_006757.3 9 UTR 5 NP_006748.1
XM_006718288.3 9 Intron XP_006718351.1
XM_006718290.3 9 Intron XP_006718353.1
XM_006718293.2 9 UTR 5 XP_006718356.1
XM_006718294.3 9 UTR 5 XP_006718357.1
XM_006718296.3 9 Intron XP_006718359.1
XM_006718299.2 9 Intron XP_006718362.1
XM_006718300.3 9 Intron XP_006718363.1
XM_006718302.3 9 Intron XP_006718365.1
XM_011520343.2 9 Intron XP_011518645.1
XM_017018205.1 9 UTR 5 XP_016873694.1
XM_017018206.1 9 Intron XP_016873695.1
XM_017018207.1 9 Intron XP_016873696.1
XM_017018208.1 9 Intron XP_016873697.1
XM_017018209.1 9 Intron XP_016873698.1
XM_017018210.1 9 Intron XP_016873699.1
XM_017018211.1 9 Intron XP_016873700.1
XM_017018212.1 9 Intron XP_016873701.1
XM_017018213.1 9 Intron XP_016873702.1
XM_017018214.1 9 Intron XP_016873703.1
XM_017018215.1 9 Intron XP_016873704.1
XM_017018216.1 9 Intron XP_016873705.1
XM_017018217.1 9 Intron XP_016873706.1
XM_017018218.1 9 Intron XP_016873707.1
XM_017018219.1 9 Intron XP_016873708.1

View Full Product Details