Product Details

SNP ID
rs755428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45702881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGGATAATCACAGAAGGATGTA[A/G]CCTATCAGTGTTAGCCAGAATAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01055 PubMed Links

Gene Details

Gene
LINC01055
Gene Name
long intergenic non-protein coding RNA 1055
There are no transcripts associated with this gene.

Gene
SPERT
Gene Name
spermatid associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286341.1 Intron NP_001273270.1
NM_001286342.1 Intron NP_001273271.1
NM_152719.2 Intron NP_689932.1
XM_011534971.2 Intron XP_011533273.1

View Full Product Details