Product Details

SNP ID
rs891186
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:55133074 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAAAATATTAGCCCTCCCTGCCCA[A/G]AGGAAGCAGTGAGCAAAACAACTGT
Phenotype
MIM: 191041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7975 PubMed Links

Gene Details

Gene
MIR7975
Gene Name
microRNA 7975
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126132.2 Intron NP_001119604.1
NM_001126133.2 Intron NP_001119605.1
NM_001291774.1 Intron NP_001278703.1
NM_003283.5 Intron NP_003274.3
XM_011527246.2 Intron XP_011525548.1
XM_017027186.1 Intron XP_016882675.1
XM_017027187.1 Intron XP_016882676.1

View Full Product Details