Product Details

SNP ID
rs919390
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41966898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGACAGGGGCGGTCCTGGGCCTG[C/G]GGGACGGGGAAGAGATGGGCGATGT
Phenotype
MIM: 182350 MIM: 604925
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256213.1 3320 UTR 3 NP_001243142.1
NM_001256214.1 3320 UTR 3 NP_001243143.1
NM_152296.4 3320 UTR 3 NP_689509.1
Gene
RABAC1
Gene Name
Rab acceptor 1
There are no transcripts associated with this gene.

View Full Product Details