Product Details

SNP ID
rs1802980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136440989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGAGCCATCATGTCAGACAGC[A/C]GGCTCTCTGGGGCTCCTTCCCAGTG
Phenotype
MIM: 613037 MIM: 612854
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
There are no transcripts associated with this gene.

Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276418.1 7994 UTR 3 NP_001263347.1
NM_014866.1 7994 UTR 3 NP_055681.1
XM_005266134.2 7994 UTR 3 XP_005266191.1
XM_011519246.2 7994 UTR 3 XP_011517548.1
XM_011519247.1 7994 UTR 3 XP_011517549.1
XM_011519248.2 7994 UTR 3 XP_011517550.1
XM_011519249.1 7994 UTR 3 XP_011517551.1
XM_011519250.2 7994 Intron XP_011517552.1
XM_011519251.1 7994 UTR 3 XP_011517553.1
XM_011519252.2 7994 UTR 3 XP_011517554.1
XM_011519253.1 7994 UTR 3 XP_011517555.1
XM_011519254.1 7994 UTR 3 XP_011517556.1
XM_011519255.2 7994 UTR 3 XP_011517557.1
XM_011519256.1 7994 UTR 3 XP_011517558.1
XM_011519257.1 7994 UTR 3 XP_011517559.1
XM_011519258.1 7994 UTR 3 XP_011517560.1
XM_011519259.1 7994 UTR 3 XP_011517561.1
XM_011519260.2 7994 UTR 3 XP_011517562.1
XM_011519261.1 7994 UTR 3 XP_011517563.1
XM_011519262.1 7994 UTR 3 XP_011517564.1
XM_011519263.2 7994 UTR 3 XP_011517565.1
XM_011519264.2 7994 Intron XP_011517566.1
XM_017015363.1 7994 UTR 3 XP_016870852.1
XM_017015364.1 7994 Intron XP_016870853.1

View Full Product Details