Product Details
- SNP ID
-
rs1436728
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:52654325 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTAGACAGCTATGACTTTTAATT[G/T]ACATTTGTTATTGTCGTGTTAGACC
- Phenotype
-
MIM: 604098
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SUGT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79027742] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUGT1
- Gene Name
- SGT1 homolog, MIS12 kinetochore complex assembly cochaperone
View Full Product Details