Product Details

SNP ID
rs1653588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121238863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTATTGCAAACTAACCAAGCCCCA[A/T]CTGGTGCCAGAAGGGGTGGAGCTAT
Phenotype
MIM: 615002 MIM: 600846
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAMKK2 PubMed Links

Gene Details

Gene
CAMKK2
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270485.1 3616 UTR 3 NP_001257414.1
NM_001270486.1 3616 UTR 3 NP_001257415.1
NM_006549.3 3616 UTR 3 NP_006540.3
NM_153499.2 3616 UTR 3 NP_705719.2
NM_153500.1 3616 UTR 3 NP_705720.1
NM_172214.2 3616 Intron NP_757363.1
NM_172215.2 3616 Intron NP_757364.1
NM_172216.1 3616 UTR 3 NP_757365.1
NM_172226.2 3616 UTR 3 NP_757380.1
XM_005253822.3 3616 UTR 3 XP_005253879.1
XM_005253823.1 3616 Intron XP_005253880.1
XM_005253824.3 3616 UTR 3 XP_005253881.1
XM_011537763.1 3616 UTR 3 XP_011536065.1
XM_011537764.2 3616 UTR 3 XP_011536066.1
XM_017018694.1 3616 UTR 3 XP_016874183.1
XM_017018695.1 3616 UTR 3 XP_016874184.1
XM_017018696.1 3616 UTR 3 XP_016874185.1
XM_017018697.1 3616 UTR 3 XP_016874186.1
XM_017018698.1 3616 UTR 3 XP_016874187.1
XM_017018699.1 3616 UTR 3 XP_016874188.1
XM_017018700.1 3616 UTR 3 XP_016874189.1
XM_017018701.1 3616 UTR 3 XP_016874190.1
XM_017018702.1 3616 UTR 3 XP_016874191.1
XM_017018703.1 3616 Intron XP_016874192.1
XM_017018704.1 3616 Intron XP_016874193.1
XM_017018705.1 3616 UTR 3 XP_016874194.1
XM_017018706.1 3616 UTR 3 XP_016874195.1
XM_017018707.1 3616 UTR 3 XP_016874196.1
XM_017018708.1 3616 Intron XP_016874197.1
XM_017018709.1 3616 UTR 3 XP_016874198.1
Gene
P2RX4
Gene Name
purinergic receptor P2X 4
There are no transcripts associated with this gene.

View Full Product Details