Product Details

SNP ID
rs1644749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47130804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCGCGGACGAGGTACTGCGCA[C/G]GCGTGGGAGGGTCTGCGCATGCGCA
Phenotype
MIM: 613294
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SAE1 PubMed Links

Gene Details

Gene
SAE1
Gene Name
SUMO1 activating enzyme subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145713.1 Intron NP_001139185.1
NM_001145714.1 Intron NP_001139186.1
NM_005500.2 Intron NP_005491.1
XM_006722963.2 Intron XP_006723026.1
XM_006722965.3 Intron XP_006723028.1
XM_017026135.1 Intron XP_016881624.1
XM_017026136.1 Intron XP_016881625.1

View Full Product Details