Product Details

SNP ID
rs1060288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28879606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACCCCATCACTCTGGCCAGGAA[A/G]AAAGATGGCACAAGGGCTCTGGGGT
Phenotype
MIM: 131560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLOT2 PubMed Links

Gene Details

Gene
FLOT2
Gene Name
flotillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004475.2 2299 UTR 3 NP_004466.2
XM_005257950.3 2299 UTR 3 XP_005258007.1
XM_005257953.2 2299 UTR 3 XP_005258010.1
XM_017024394.1 2299 UTR 3 XP_016879883.1
XM_017024395.1 2299 UTR 3 XP_016879884.1
XM_017024396.1 2299 UTR 3 XP_016879885.1

View Full Product Details