Product Details

SNP ID
rs1044813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90231027 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCAGACTCCTCCAGGATCTGG[C/G]AAAGGGAGAGTTTCAGGCCAGAGCC
Phenotype
MIM: 602293 MIM: 617029
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIB1 PubMed Links

Gene Details

Gene
CIB1
Gene Name
calcium and integrin binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277764.1 Intron NP_001264693.1
NM_006384.3 Intron NP_006375.2
XM_006720375.1 Intron XP_006720438.1
Gene
GDPGP1
Gene Name
GDP-D-glucose phosphorylase 1
There are no transcripts associated with this gene.

Gene
SEMA4B
Gene Name
semaphorin 4B
There are no transcripts associated with this gene.

View Full Product Details