Product Details

SNP ID
rs894037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104927186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCAGTGGCCCCCACATGGCCA[C/T]GCTCCATGCCGCCCCGCCGCCCGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLD4 PubMed Links

Gene Details

Gene
PLD4
Gene Name
phospholipase D family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308174.1 221 Missense Mutation CGC,TGC R23C NP_001295103.1
NM_138790.3 221 Missense Mutation CGC,TGC R16C NP_620145.2
XM_011536411.2 221 Missense Mutation CGC,TGC R23C XP_011534713.1
XM_017020965.1 221 Missense Mutation CGC,TGC R16C XP_016876454.1

View Full Product Details