Product Details
- SNP ID
-
rs1047928
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:3736053 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCCGTGTTTTTCGTGTAAGATC[C/G]TGCTTCTCCGAGCAGTATTAATGGA
- Phenotype
-
MIM: 601990
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TP73
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80098330] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TP73
- Gene Name
- tumor protein p73
- Gene
- TP73-AS1
- Gene Name
- TP73 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details