Product Details
- SNP ID
-
rs3903158
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:3738972 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGCAATTTTCCACCTGAAATTGG[C/G]ATATCATCTGCACTGACTGCAGCCT
- Phenotype
-
MIM: 601990
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TP73
PubMed Links
Gene Details
- Gene
- TP73
- Gene Name
- tumor protein p73
There are no transcripts associated with this gene.
- Gene
- TP73-AS1
- Gene Name
- TP73 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details