Product Details

SNP ID
rs930569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:86088113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACATGCAAATGAATGGGTATGC[A/C]TGTGTTCCAGTGGAACTTTATTTGG
Phenotype
MIM: 615496
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGBL1 PubMed Links

Gene Details

Gene
AGBL1
Gene Name
ATP/GTP binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011521226.2 Intron XP_011519528.1
XM_011521227.2 Intron XP_011519529.1
XM_011521228.2 Intron XP_011519530.1
XM_011521230.2 Intron XP_011519532.1
XM_017021918.1 Intron XP_016877407.1
XM_017021919.1 Intron XP_016877408.1
XM_017021920.1 Intron XP_016877409.1
XM_017021921.1 Intron XP_016877410.1
Gene
LINC01584
Gene Name
long intergenic non-protein coding RNA 1584
There are no transcripts associated with this gene.

View Full Product Details