Product Details

SNP ID
rs1057946
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:84639555 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTGAACTCCCACGTGGAGTAGGAT[G/T]AAAACCCACAGCATCTAGATCTTCA
Phenotype
MIM: 610417 MIM: 616144
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCAND2P PubMed Links

Gene Details

Gene
SCAND2P
Gene Name
SCAN domain containing 2 pseudogene
There are no transcripts associated with this gene.

Gene
WDR73
Gene Name
WD repeat domain 73
There are no transcripts associated with this gene.

View Full Product Details