Product Details

SNP ID
rs905938
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:155018913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCCACGAGGCGTACCAGCCCA[C/T]GGCTGTAGATATTTTCTGATTTTAG
Phenotype
MIM: 607646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCST2 PubMed Links

Gene Details

Gene
DCST2
Gene Name
DC-STAMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144622.2 Intron NP_653223.2
XM_011509188.2 Intron XP_011507490.1
XM_011509189.2 Intron XP_011507491.1
Gene
ZBTB7B
Gene Name
zinc finger and BTB domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252406.2 Intron NP_001239335.1
NM_001256455.1 Intron NP_001243384.1
XM_006711349.2 Intron XP_006711412.2
XM_006711353.1 Intron XP_006711416.1
XM_006711354.1 Intron XP_006711417.1
XM_006711356.3 Intron XP_006711419.1
XM_006711357.1 Intron XP_006711420.1
XM_006711358.1 Intron XP_006711421.1
XM_006711359.2 Intron XP_006711422.1
XM_011509598.2 Intron XP_011507900.1
XM_011509599.2 Intron XP_011507901.1
XM_017001399.1 Intron XP_016856888.1

View Full Product Details