Product Details

SNP ID
rs9874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101271199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACATTTTCAGCTACAGAAAATA[C/T]ATTCAAGGAAGAATGTTAAAAAGTT
Phenotype
MIM: 607918
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VIMP PubMed Links

Gene Details

Gene
VIMP
Gene Name
VCP interacting membrane selenoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018445.5 1127 Intron NP_060915.2
NM_203472.2 1127 UTR 3 NP_982298.2

View Full Product Details