Product Details

SNP ID
rs3893462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7493286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGGTGCTTATAGTTATAGGGA[A/G]GACAGACCCCCATAGGTCATTTTAT
Phenotype
MIM: 180660 MIM: 612308
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR2A PubMed Links

Gene Details

Gene
POLR2A
Gene Name
RNA polymerase II subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000937.4 Intron NP_000928.1
Gene
ZBTB4
Gene Name
zinc finger and BTB domain containing 4
There are no transcripts associated with this gene.

View Full Product Details